Learn More
B Braun™ Durogrip™ Needle Holders
Product Code. 12678233
Change view
Click to view available options
Length (Metric):
125 mm
145 mm
Style:
Hegar-Olsen
Webster
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Style Unit Size
12678233 145 mm Hegar-Olsen Each
10433315 125 mm Webster Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12678233 Supplier B Braun™ Supplier No. BM128R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
An instrument for guiding needles, suturing and gripping fine or course tissues.
  • Serrated jaws on both varieties
  • Jaws with tungsten carbide inserts
  • Safe hold on needle
Specifications
Product Type Needle holders
Description Serrated jaws
For Use With (Application) Guiding/holding needles and sutures
Style Hegar-Olsen
Length (Metric) 145 mm