Learn More
B Braun™ Durogrip™ Needle Holders
An instrument for guiding needles, suturing and gripping fine or course tissues.
€ 160.00 - € 168.00
Specifications
Product Type | Needle holders |
---|---|
Description | Serrated jaws |
For Use With (Application) | Guiding/holding needles and sutures |
Product Code | Brand | Style | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Style | Length (Metric) | Price | Quantity & Availability | ||||
10433315
|
B Braun™
BM024R |
Webster | 125 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12678233
|
B Braun™
BM128R |
Hegar-Olsen | 145 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Serrated jaws on both varieties
- Jaws with tungsten carbide inserts
- Safe hold on needle
Specifications
Needle holders | |
Serrated jaws | |
Guiding/holding needles and sutures |