missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP10 Monoclonal Antibody (OTI2E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806723S
This item is not returnable.
View return policy
Description
DUSP10 Monoclonal antibody specifically detects DUSP10 in Human samples. It is validated for Western BlotSpecifications
| DUSP10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P10451 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6696 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI2E3 | |
| Unconjugated | |
| SPP1 | |
| 2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase | |
| Human recombinant protein fragment corresponding to amino acids 17-314 of human SPP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction