missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106794
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DUSP10.
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
Specifications
| DUSP10 | |
| Polyclonal | |
| Unconjugated | |
| DUSP10 | |
| 2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11221, 63953, 63995 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ESS0, Q9Y6W6 | |
| DUSP10 | |
| A synthesized peptide derived from human DUSP10(Accession Q9Y6W6), corresponding to amino acid residues H357-H407. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction