missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP10 Polyclonal Antibody

Description
Immunogen sequence: FMEYNKSHIQ GAVHINCADK ISRRRLQQGK ITVLDLISCR EGKDSFKRIF SKEIIVYDEN TNEPSRVMPS QPLHIVLESL KREGKEPLVL KGGLSSFKQN HENLCDNSLQ LQ Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. GPSN2 catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA. Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2, 3-enoyl-CoA to saturated acyl-CoA.
Specifications
Specifications
| Antigen | DUSP10 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DUSP10 |
| Gene Accession No. | Q9Y6W6 |
| Gene Alias | 2610306G15Rik; AI158871; dual specificity phosphatase 10; dual specificity phosphatase MKP-5; dual specificity protein phosphatase 10; Dusp10; MAP kinase phosphatase 5; mitogen-activated protein kinase phosphatase 5; Mkp5; MKP-5; serine/threonine specific protein phosphatase |
| Gene Symbols | DUSP10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?