missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP12 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18420C
This item is not returnable.
View return policy
Description
DUSP12 Polyclonal antibody specifically detects DUSP12 in Human samples. It is validated for Western Blot
Specifications
| DUSP12 | |
| Polyclonal | |
| Western Blot | |
| dual specificity phosphatase 12, dual specificity protein phosphatase 12, Dual specificity tyrosine phosphatase YVH1, DUSP1YVH1 protein-tyrosine phosphatase ortholog, EC 3.1.3.48, serine/threonine specific protein phosphatase, TDSP4, YVH1EC 3.1.3.16 | |
| Recognizes region between residues Between Between 1 and 50.. | |
| 0.1 mL | |
| Protein Phosphatase | |
| 11266 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction