Learn More
DUSP12 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand:  Abnova H00011266-AP21.1Set
Product Code. 16163234
  • € 843.00 / 1 set
Specifications
DUSP12
Unconjugated
DUSP1/YVH1
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-DUSP12 (100 ug) 2. Detection antibody: mouse purified polyclonal anti-DUSP12 (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
dual specificity phosphatase 12
DUSP12
RUO
11266
Human
Ab Pair