missing translation for 'onlineSavingsMsg'
Learn More

DUSP14 Antibody [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30488242 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488242 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488242 Supplier Novus Biologicals Supplier No. NBP335121V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DUSP14 Polyclonal antibody specifically detects DUSP14 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP14
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias dual specificity phosphatase 14, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 6, Mitogen-activated protein kinase phosphatase 6, MKP-1 like protein tyrosine phosphatase, MKP-1-like protein tyrosine phosphatase, MKP-6, MKP6MKP-Ldual specificity protein phosphatase 14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-198 of human DUSP14 (NP_008957.1).,, Sequence:, MSSRGHSTLPRTLMAPRMISEGDIGGIAQITSSLFLGRGSVASNRHLLQARGITCIVNATIEIPNFNWPQFEYVKVPLADMPHAPIGLYFDTVADKIHSVSRKHGATLVHCAAGVSRSATLCIAYLMKFHNVCLLEAYNWVKARRPVIRPNVGFWRQLIDYERQLFGKSTVKMVQTPYGIVPDVYEKESRHLMPYWGI
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 11072
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.