missing translation for 'onlineSavingsMsg'
Learn More

DUSP15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15829233
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15829233 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829233 Supplier Invitrogen Supplier No. PA570822

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DUSP15 Polyclonal Antibody

DUSP15 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP15
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dusp15
Gene Accession No. Q8R4V2
Gene Alias AI851682; C20orf57; dual specificity phosphatase 15; dual specificity phosphatase-like 15; dual specificity protein phosphatase 15; Dual specificity protein phosphatase T-DSP10; DUSP15; FLJ40111; LMW-DSP10; LOW QUALITY PROTEIN: dual specificity protein phosphatase 15; RP11-243J16.11; T-DSP10; Vaccinia virus VH1-related dual-specific protein phosphatase Y; VH1-related member Y; VHY
Gene Symbols Dusp15
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse DUSP15.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 252864
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.