missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP23 Antibody (OTI3C10), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71976MFV450
This item is not returnable.
View return policy
Description
DUSP23 Monoclonal antibody specifically detects DUSP23 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| DUSP23 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3C10 | |
| Flow Cytometry | |
| dual specificity phosphatase 23, dual specificity protein phosphatase 23, DUSP25, EC 3.1.3.16, EC 3.1.3.48, FLJ20442, LDP3, LDP-3, Low molecular mass dual specificity phosphatase 3, low-molecular-mass dual-specificity phosphatase 3, MOSP, RP11-190A12.1, VH1-like member Z, VH1-like phosphatase Z, VHZ | |
| Full length human recombinant protein of human DUSP23(NP_060293) produced in HEK293T cell. | |
| 0.1 mL | |
| Protein Phosphatase | |
| 54935 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction