missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP5 Monoclonal Antibody (OTI5D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811035S
This item is not returnable.
View return policy
Description
DUSP5 Monoclonal antibody specifically detects DUSP5 in Human samples. It is validated for Western BlotSpecifications
| DUSP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16690 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1847 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5D10 | |
| Unconjugated | |
| DUSP5 | |
| candidate plasticity-related gene 21; Cpg21; dual specificity phosphatase 5; dual specificity protein phosphatase 5; dual specificity protein phosphatase 5-like; dual specificity protein phosphatase hVH3; DUSP; DUSP5; Gm337; HVH3; LOW QUALITY PROTEIN: dual specificity protein phosphatase 5; MAP-kinase phosphatase (cpg21); MAP-kinase phosphatase CPG21; serine/threonine specific protein phosphatase; VH1-like phosphatase 3; VH3 | |
| Human recombinant protein fragment corresponding to amino acids 125-384 of human DUSP5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction