missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585961
This item is not returnable.
View return policy
Description
DUSP5 Polyclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein kinase superfamily, which are associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates ERK1, is expressed in a variety of tissues with the highest levels in pancreas and brain, and is localized in the nucleus.
Specifications
| DUSP5 | |
| Polyclonal | |
| Unconjugated | |
| DUSP5 | |
| candidate plasticity-related gene 21; Cpg21; dual specificity phosphatase 5; dual specificity protein phosphatase 5; dual specificity protein phosphatase 5-like; dual specificity protein phosphatase hVH3; DUSP; DUSP5; Gm337; HVH3; LOW QUALITY PROTEIN: dual specificity protein phosphatase 5; MAP-kinase phosphatase (cpg21); MAP-kinase phosphatase CPG21; serine/threonine specific protein phosphatase; VH1-like phosphatase 3; VH3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1847, 240672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q16690 | |
| DUSP5 | |
| Synthetic peptide within Human DUSP5 aa 71-120. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto