missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548932
This item is not returnable.
View return policy
Description
DUX3 Polyclonal Antibody for Western Blot
The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX3, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM606009).
Specifications
| DUX3 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96PT4 | |
| DUX3 | |
| KLH conjugated synthetic peptide between 21-48 amino acids from the N-terminal region of human DUX3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DUX3 | |
| double homeobox 3; double homeobox, 3; DUX3; putative double homeobox protein 3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 26582 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction