missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | DVL1 |
|---|---|
| Clone | 1A2A5 |
| Concentration | 1100 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237895
|
Proteintech
67672-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236721
|
Proteintech
67672-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DVL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DVL1. This antibody recognizes human, rat, mouse antigen. The DVL1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| DVL1 | |
| 1100 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 1855 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
| 1A2A5 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Mouse | |
| DVL; MGC54245 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title