missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL2 Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806790
This item is not returnable.
View return policy
Description
Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.Specifications
| DVL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14641 | |
| DVL2 | |
| Human recombit protein fragment corresponding to amino acids 1-320 of human DVL2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E11 | |
| Unconjugated | |
| DVL2 | |
| dishevelled segment polarity protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1856 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction