missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL2 Monoclonal Antibody (OTI1E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806790S
This item is not returnable.
View return policy
Description
DVL2 Monoclonal antibody specifically detects DVL2 in Human samples. It is validated for Western BlotSpecifications
| DVL2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14641 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1856 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1E11 | |
| Unconjugated | |
| DVL2 | |
| dishevelled 2; dishevelled 2 (homologous to Drosophila dsh); dishevelled 2, dsh homolog; dishevelled 2, dsh homolog (Drosophila); dishevelled segment polarity protein 2; dishevelled, dsh homolog 2; dishevelled-2; DSH homolog 2; Dvl2; Dvl-2; LOW QUALITY PROTEIN: segment polarity protein dishevelled homolog DVL-2; sb:eu471; Segment polarity protein dishevelled homolog DVL-2; wu:fc05d12; wu:fo71e09; wu:fp54a02; zgc:55372 | |
| Human recombinant protein fragment corresponding to amino acids 1-320 of human DVL2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction