Learn More
DVL2 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12037-100UL
 View more versions of this product
Product Code. 18057315
  • € 450.00 / 100µL
Description
The DVL2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human DVL2. This antibody recognizes human, mouse, rat antigen. The DVL2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
DVL2
1000 μg/mL
Dishevelled 2, DSH homolog 2, DVL2
100 μL
1856
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa