missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DXD Antibody (PSH0-36), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-33117
This item is not returnable.
View return policy
Description
DXD Monoclonal antibody specifically detects DXD in Nonspecies specific samples. It is validated for ELISA
Specifications
| DXD | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:1000 | |
| 1599440-33-1, Exatecan derivative for ADC | |
| DXD-OVA | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| ELISA | |
| PSH0-36 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction