missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558107
This item is not returnable.
View return policy
Description
Immunogen sequence: ELEMQEKERQ RIQELQRAQE QLGKEMRMNM ENLVRNEDIL HSEEATLDSG KTLAEISDRY GAPNLSRVEE LDEPMFSDI Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
This gene encodes a member of a family of proteins that contains a DPY30 domain. The encoded protein is involved in acrosome formation during spermatid development. This gene locus overlaps with a closely related gene on the opposite strand. Alternative splicing results in multiple transcript variants.
Specifications
| DYDC1 | |
| Polyclonal | |
| Unconjugated | |
| DYDC1 | |
| 1700029M23Rik; bA36D19.5; BC019250 protein; DPY30 domain containing 1; DPY30 domain-containing protein 1; DPY30D1; DYDC1; FLJ43920; OTTHUMP00000019963; RSD9; RSD-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 143241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWB3 | |
| DYDC1 | |
| Recombinant protein corresponding to Human DYDC1. Recombinant protein control fragment (Product #RP-97167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction