missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00214JF646
This item is not returnable.
View return policy
Description
DYDC2 Polyclonal antibody specifically detects DYDC2 in Human samples. It is validated for ELISA
Specifications
| DYDC2 | |
| Polyclonal | |
| ELISA | |
| bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
| Produced in rabbits immunized with purified, recombinant Human DYDC2 (Uniprot#: Q96IM9; Thr102-Pro176) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84332 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction