missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC2 Rabbit anti-Human, Clone: 101, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 367.00 - € 552.00
Specifications
| Antigen | DYDC2 |
|---|---|
| Clone | 101 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18207937
|
Novus Biologicals
NBP2-90258-50UL |
50 μg |
€ 367.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203896
|
Novus Biologicals
NBP2-90258-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DYDC2 Monoclonal specifically detects DYDC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| DYDC2 | |
| Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 84332 | |
| IgG | |
| Protein A purified |
| 101 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
| Recombinant Human DYDC2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title