missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYKDDDDK Mouse anti-Recombinant Protein, CoraLite 488, Clone: 1E7B4, Proteintech
Shop All Proteintech ProductsDescription
The DYKDDDDK antibody from Proteintech is a mouse monoclonal antibody generated with peptide of DYKDDDDK. This antibody recognizes recombinant protein antigen. The DYKDDDDK antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
| Antigen | DYKDDDDK |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1E7B4 |
| Conjugate | CoraLite 488 |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Isotype | IgG2b |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?