missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ FlowComp™ Human CD8 Kit

Product Code. 10206403
Change view
Click to view available options
Quantity:
3 mL
Unit Size:
3mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10206403 3 mL 3mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10206403 Supplier Invitrogen™ Supplier No. 11362D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This gentle and easy-to-use kit allows you to isolate flow-compatible human CD8+ T-cells from PBMC or directly from whole blood.

This gentle and easy-to-use kit allows you to isolate flow-compatible human CD8+ T-cells from PBMC or directly from whole blood. Immediately after positive isolation, the beads are released and removed from your cell sample. In this way, you avoid exposing your cells to potentially cytotoxic or immunogenic foreign substances like iron oxides or dextrans, a potential problem when working with biodegradable particles. In addition, your cells will not be exposed to the stress of being passed through a column. You can expect a purity level of >95% human CD8+ cells.

Advantages:
• No risk of your results being affected by the isolation method
• Gentle tube-based method - no columns needed
• High purity and recovery
• Bead-free cells - compatible with flow cytometry analysis
• Compatible with flow cytometry analysis
• Viable, bead-free CD8+ T-cells for direct use in any cell-based assay.

The isolated cells have the presumed profile of memory and naive CD8+ T cells, and show a high proliferation capacity. You will save time and money, and get increased confidence in your results.

Starting sample:
MNC or whole blood.

Capacity:
The kit will process 2 × 109 mononuclear cells (MNC) or 80 mL whole blood

TRUSTED_SUSTAINABILITY

Specifications

Final Product Type Human CD8+ T cells
Product Line Dynabeads, FlowComp
Product Type Human CD8 Isolation Kit
Quantity 3 mL
Sufficient For 2 x 109 cells
Content And Storage The Kit Contains:
• 1 mL Antibody
• 3 mL FlowComp™ Dynabeads™
• 40 mL FlowComp™ Release Buffer.

Store at: 2°C to 8°C
Capacity Processes ∼2 x 109 cells total
Stationary Phase PBMC, Blood
Purity or Quality Grade Research Grade
Shipping Condition Room Temperature
Species Human
Matrix Magnetic Beads
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.