missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynactin 6 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97258FR
This item is not returnable.
View return policy
Description
Dynactin 6 Polyclonal antibody specifically detects Dynactin 6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Dynactin 6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| dynactin 6, dynactin subunit 6, Dynactin subunit p27, P27, Protein WS-3, WS-3, WS3novel RGD-containing protein | |
| Produced in rabbits immunized with E. coli-derived Human Dynactin 6 fragment. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Immunology, Signal Transduction | |
| 10671 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction