Learn More
Dynactin Subunit 1/DCTN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-21351-25ul
 View more versions of this product
Product Code. 18656374
  • € 401.00 / 25µL
Description
Dynactin Subunit 1/DCTN1 Polyclonal antibody specifically detects Dynactin Subunit 1/DCTN1 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dynactin Subunit 1/DCTN1
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), p135, p150 Glued (Drosophila) homolog, p150-glued
This antibody was developed against Recombinant Protein corresponding to amino acids: ALYRKTSQLLETLNQLSTHTHVVDITRTSPAAKSPSAQLMEQVAQLKSLSDTVEKLKDEVLKETVSQRPGATV
25 μg
Cell Cycle and Replication
1639
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol