missing translation for 'onlineSavingsMsg'
Learn More

Dynactin Subunit 1/DCTN1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-71829PE

Product Code. 30035265

  • € 615.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Dynactin Subunit 1/DCTN1 Polyclonal antibody specifically detects Dynactin Subunit 1/DCTN1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dynactin Subunit 1/DCTN1
Polyclonal
Western Blot, Immunoprecipitation
dynactin 1, dynactin 1 (p150, Glued (Drosophila) homolog), dynactin subunit 1, glued homolog, Drosophila), p135, p150 Glued (Drosophila) homolog, p150-glued
The immunogen for this product maps to a region between residue 1228 to 1278 of human 150 kDa Dynein-Associated Polypeptide using the numbering given in entry NP_004073.2 (GeneID 1639).
0.1 mL
Cell Cycle and Replication
1639
Store at 4C in the dark.
IgG
Western Blot, Immunoprecipitation
PE
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.