Learn More
Abeba™ Dynamic 7345 Shoes
Product Code. 15602778
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15602778
Brand: Abeba™ 7345/48
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Microfiber with breathable textile inlays
  • White/Black
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3574)
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRC
Color White, Black
Material Microfiber
Size 48
Sole Features Slip-resistant
Sole Material Polyurethane
Style 7345