missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | Dynamin 3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Dynamin 3 Polyclonal antibody specifically detects Dynamin 3 in Human, Mouse samples. It is validated for Western BlotSpecifications
| Dynamin 3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS with 50% glycerol, pH7.3. | |
| 26052 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Dyna III, dynamin 3, dynamin family member, dynamin-3, EC 3.6.5.5, MGC70433, T-dynamin, testicular | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 594-863 of human Dynamin 3 (NP_056384.2). KDYRFLELACDSQEDVDSWKASLLRAGVYPDKSVGNNKAENDENGQAENFSMDPQLERQVETIRNLVDSYMSIINKCIRDLIPKTIMHLMINNVKDFINSELLAQLYSSEDQNTLMEESAEQAQRRDEMLRMYQALKEALGIIGDISTATVSTPAPPPVDDSWIQHSRRSPPPSPTTQRRPTLSAPLARPTSGRGPAPAIPSPGPHSGAPPVPFRPGPLPPFPSSSDSFGAPPQVPSRPTRAPPSVPSRRPPPSPTRPTIIRPLESSLLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title