missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynamin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00
Specifications
| Antigen | Dynamin |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698735
|
Novus Biologicals
NBP2-48950-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648285
|
Novus Biologicals
NBP2-48950 |
0.1 mL |
N/A
|
N/A | |||||
Description
Dynamin Polyclonal antibody specifically detects Dynamin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Dynamin | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Membrane Trafficking and Chaperones, Neuroscience, Signal Transduction | |
| PBS (pH 7.2), 40% Glycerol | |
| 1759 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DNM, dynamin 1, dynamin-1, EC 3.6.5.5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: AGVYPERVGDKEKASETEENGSDS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title