missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYNC1I2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12219-1-AP-20UL
This item is not returnable.
View return policy
Description
Dyneins are multi-subunit, high molecular weight ATPases that interact with microtubules to generate force by converting the chemical energy of ATP into the mechanical energy of movement. Cytoplasmic or axonemal Dynein heavy, intermediate, light and light-intermediate chains are all components of minus end-directed motors; the complex that transports cellular cargos towards the central region of the cell. Axonemal Dynein motors contain one to three non-identical heavy chains and cause a sliding of microtubules in the axonemes of cilia and flagella in a mechanism necessary for cilia to beat and propel the cell. Cytoplasmic Dynein is an approximately 12 subunit complex of two heavy chains, two intermediate chains to anchor Dynein to its cargo, four smaller intermediate chains and several light chains. It performs functions necessary for cell survival such as organelle transport and centrosome assembly. The carboxy-terminus of Dynein is important for microtubule-dependent motility and is highly conserved, while the amino-terminal regions are more variable. Several proteins regulate Dynein activity, including Dynactin, LIS1 and nudel (nudE-like).Specifications
| DYNC1I2 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1I2 | |
| DH IC 2, DNCI2, DNCIC2, DYNC1I2, IC2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116659, 13427, 1781 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88487, Q13409, Q62871 | |
| DYNC1I2 | |
| DYNC1I2 Fusion Protein Ag2873 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction