missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC1LI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578491
This item is not returnable.
View return policy
Description
Positive Control: 293T, HepG2, mouse brain Predicted Reactivity: Rat (91%), Xenopus laevis (83%), Chicken (87%), Chimpanzee (99%), Bovine (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S12E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal cancers compared to matched normal colonic mucosa has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| DYNC1LI1 | |
| Polyclonal | |
| Unconjugated | |
| DYNC1LI1 | |
| 1110053F02Rik; Cytoplasmic dynein 1 light intermediate chain 1; DLC-A; DNCLI1; Dnclic1; Dync1li1; dynein cytoplasmic 1 light intermediate chain 1; dynein light chain A; Dynein light intermediate chain 1, cytosolic; dynein, cytoplasmic, light intermediate chain 1; dynein, cytoplasmic, light intermediate polypeptide 1; LIC1; LIC-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 235661, 51143 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8R1Q8, Q9Y6G9 | |
| DYNC1LI1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human DYNC1LI1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction