missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYNC2LI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106406
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DYNC2LI1.
DYNC2LI1 (dynein, cytoplasmic 2, light intermediate chain 1), also known as LIC3, D2LIC or CGI-60, is a 351 amino acid cytoplasmic protein belonging to the dynein light intermediate chain family. DYNC2LI1 may function as a motor for intraflagellar retrograde transport and in cilia biogenesis. The cytoplasmic dynein complex 2 may be composed of a DYNC2H1 homodimer and a number of DYNC2LI1 light intermediate chains. DYNC2LI1 exists as five alternatively spliced isoforms.
Specifications
| DYNC2LI1 | |
| Polyclonal | |
| Unconjugated | |
| Dync2li1 | |
| 4933404O11Rik; CGI-60; Cytoplasmic dynein 2 light intermediate chain 1; D2LIC; Dync2li1; Dynein 2 light intermediate chain; dynein cytoplasmic 2 light intermediate chain 1; dynein, cytoplasmic 2, light intermediate chain 1; LIC3; mD2LIC; RGD1310286 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 213575, 298767, 51626 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AY43, Q8K0T2, Q8TCX1 | |
| Dync2li1 | |
| A synthesized peptide derived from human DYNC2LI1(Accession Q8TCX1), corresponding to amino acid residues R54-D104. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction