missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dynein light chain 4 Antibody (3B7B7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37390
This item is not returnable.
View return policy
Description
Dynein light chain 4 Monoclonal specifically detects Dynein light chain 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| Dynein light chain 4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| dJ327J16, dynein light chain 4, axonemal, dynein light chain, outer arm 4, dynein, axonemal, light 4, dynein, axonemal, light chain 4, dynein, axonemal, light polypeptide 4, PIG27, proliferation-inducing gene 27, proliferation-inducing protein 27 | |
| Mouse | |
| 12 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 3B7B7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DNAL4 | |
| Purified recombinant fragment of human DNAL4 (AA: 1-105) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| 10126 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction