Learn More
JULABO™ DYNEO™ DD-201F Refrigerated/Heating Circulator
Product Code. 15900277
Click to view available options
Includes:
Analog interface
Interface not included
RS232 interface
Unit Size:
Each
Product Code. 15900277
Brand: JULABO™ 9021702.A
Only null left
Add to basket
Add to basket
Circulators have a wide working temperature range. Suitable for both internal and external applications. Pump capacity is 22 L/min with pressure of 0.6 bar. The cooling machines operate precisely and reliably even at elevated ambient temperatures up to +40 °C.
  • Powerful cooling machines
  • Suitable for internal and external applications
  • Optimized cooling coil design saves space in the bath tank
  • Powerful and infinitely adjustable pressure pump
  • Flow rate 22 L/min, pressure 0.6 bar
  • Easy switching between internal and external circulation
  • Large color TFT display, multilingual interface
  • Central rotary knob (controller) simplifies operation
  • Integrated programmer for 8 x 60 program steps
  • Integrated external Pt100 connection
  • USB port
  • RS232 interface or analog interfaces (optional)
  • Bath cover included with delivery
  • Integrated drain makes emptying liquid easy and safe
Specifications
Description Refrigerated/Heating Circulator
Capacity (Metric) 3 to 4 L
Cooling Capacity 0.2 kW
Flow Rate 8 to 23 L/min.
Display 3.5 in. TFT Display
Heating Capacity 2 kW
Includes Analog interface
Model DYNEO™ DD-201F
Partial Flow 0.1 to 0.6 bar
Temperature (Metric) Maximum -20°C to 200°C
Dimensions (L x W x H) 44 x 41 x 44 cm
Depth (Metric) Interior 15 cm
Weight (Metric) 24.7 kg
Voltage 100 V/50-60 Hz, 115 V/60 Hz, 230 V/50-60 Hz