missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DYNLRB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15829103
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15829103 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15829103 Supplier Invitrogen™ Supplier No. PA570692

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The DYNLRB1 gene is a member of the roadblock dynein light chain family. The encoded cytoplasmic protein is capable of binding intermediate chain proteins, interacts with transforming growth factor-beta, and has been implicated in the regulation of actin modulating proteins. Upregulation of this gene has been associated with hepatocellular carcinomas, suggesting that this gene may be involved in tumor progression. Alternative splicing results in multiple transcript variants. Pseudogenes of this gene have been defined on chromosomes 12 and 18.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYNLRB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dynlrb1
Gene Accession No. Q9NP97
Gene Alias 2010012N15Rik; 2010320M17Rik; 9430076K19Rik; AV124457; BITH; bithoraxoid-like protein; BLP; Dncl2a; DNLC2A; dynein light chain 2A, cytoplasmic; Dynein light chain roadblock-type 1; dynein, cytoplasmic, light chain 2A; dynein, cytoplasmic, light polypeptide 2A; dynein, light chain, roadblock-type 1; dynein-associated protein Km23; dynein-associated protein RKM23; DYNLRB1; HSPC162; hypothetical protein LOC513342; km23; km23-1; Roadblock domain-containing protein 1; ROBL/LC7-like 1; robl/LC7-like protein; ROBLD1; similar to dynein, cytoplasmic, light polypeptide 2A; zgc:56335
Gene Symbols Dynlrb1
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human DYNLRB1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83658
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.