missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrobrevin alpha Antibody (OTI1B2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70591APC
This item is not returnable.
View return policy
Description
Dystrobrevin alpha Monoclonal specifically detects Dystrobrevin alpha in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Dystrobrevin alpha | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| DTNA | |
| Full length recombinant protein of human Dystrobrevin alpha (NP_116761) produced in HEK293T cells. | |
| 0.1 mL | |
| 1837.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1B2 | |
| Flow Cytometry | |
| Alpha-dystrobrevin, D18S892EDTN, DRP3FLJ96209, DTN-1, DTN-2, DTN-3, DTN-A, dystrobrevin alpha, dystrobrevin, alpha, Dystrophin-related protein 3, LVNC1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction