Learn More
Dystrophin Antibody (4B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26617-50ul
 View more versions of this product
Product Code. 30043449
  • € 343.00 / 50µL
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dystrophin
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
4B10
ELISA, Immunohistochemistry 1:50-1:200
BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142
A synthesized peptide derived from Human Dystrophin [UniProt P11532]
50 μL
Cell Biology, Cellular Markers
1756
Store at -20 to -70C. Avoid freeze-thaw cycles.