missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Antibody (DMD/3244), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08702R
This item is not returnable.
View return policy
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Dystrophin | |
| Monoclonal | |
| DyLight 550 | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| A recombinant fragment (around aa 114-263) of human DMD protein (exact sequence is proprietary) (Uniprot: P11532) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 1756 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Protein Array | |
| DMD/3244 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction