missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Antibody (DMD/6270), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20751APC
This item is not returnable.
View return policy
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human, Canine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Dystrophin | |
| Monoclonal | |
| APC | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| Recombinant fragment (around aa1700-2300) of human Dystrophin (exact sequence is proprietary). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 1756 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DMD/6270 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction