missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystrophin Antibody (DMD/8773R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 311.00 - € 561.00
Specifications
| Antigen | Dystrophin |
|---|---|
| Clone | DMD/8773R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662105
|
Novus Biologicals
NBP3-20476-100ug |
100 μg |
€ 561.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664724
|
Novus Biologicals
NBP3-20476-20ug |
20 μg |
€ 311.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dystrophin Monoclonal antibody specifically detects Dystrophin in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Dystrophin | |
| 0.2 mg/ml | |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| 10mM PBS with 0.05% BSA | |
| 1756 | |
| IgG κ | |
| Protein A or G purified |
| DMD/8773R | |
| Western Blot 1-2 μg/mL, Immunocytochemistry/Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BMDDXS272, CMD3B, DXS142, DXS164, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, dystrophin, dystrophin (muscular dystrophy, Duchenne and Becker types), includes DXS142 | |
| Recombinant fragment (around aa1700-2300) of human Dystrophin (exact sequence is proprietary). | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title