missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DYTN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548974
This item is not returnable.
View return policy
Description
DYTN Polyclonal Antibody for Western Blot
Specifications
| DYTN | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A2CJ06 | |
| DYTN | |
| KLH conjugated synthetic peptide between 419-447 amino acids from the C-terminal region of human DYTN. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DYTN | |
| Dystrotelin; DYTN | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 391475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction