missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYX1C1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-68815
This item is not returnable.
View return policy
Description
DYX1C1 Polyclonal antibody specifically detects DYX1C1 in Human, Canine samples. It is validated for Western Blot, Peptide ELISA
Specifications
| DYX1C1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
| Western Blot, Peptide ELISA | |
| 0.5 mg/mL | |
| Western Blot, Peptide ELISA detection limit 1:32000 | |
| dyslexia susceptibility 1 candidate 1, dyslexia susceptibility 1 candidate gene 1 protein, DYX1, DYXC1, EKN1MGC70618, FLJ37882, RD | |
| DYX1C1/ EKN1, N Terminus | |
| 0.1 mg | |
| Vision | |
| 161582 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction