Learn More
DZIP3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97549MFV610
This item is not returnable.
View return policy
Description
DZIP3 Polyclonal antibody specifically detects DZIP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DZIP3 | |
| Immunohistochemistry (Paraffin) | |
| Store at 4C in the dark. | |
| Purified |