missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DZIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532189
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431. Predicted reactivity: Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 Ubiquitin ligase proteins mediate ubiquitination and subsequent proteasomal degradation of target proteins. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Able to specifically bind RNA.
Specifications
| DZIP3 | |
| Polyclonal | |
| Unconjugated | |
| DZIP3 | |
| 2310047C04Rik; 2A-HUB; 6430549P11Rik; A230104G20; DAZ interacting protein 3, zinc finger; DAZ interacting zinc finger protein 3; DAZ-interacting protein 3; DAZ-interacting protein 3 homolog; DZIP3; E3 ubiquitin-protein ligase DZIP3; hRUL138; human RNA-binding ubiquitin ligase of 138 kDa; Kiaa0675; PPP1R66; protein phosphatase 1, regulatory subunit 66; RGD1563278; RING-type E3 ubiquitin transferase DZIP3; RNA-binding RING-H2 protein-ubiquitin ligase; RNA-binding ubiquitin ligase of 138 kDa; UURF2; UURF2 ubiquitin ligase; zinc finger DAZ interacting protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9666 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.93 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q86Y13 | |
| DZIP3 | |
| Recombinant protein encompassing a sequence within the center region of human DZIP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction