Learn More
(E)-2-Methyl-3-(4-methyl-2-oxo-2H-pyran-6-yl)propenal, TRC
Specifications
Specifications
| Chemical Name or Material | (E)-2-Methyl-3-(4-methyl-2-oxo-2H-pyran-6-yl)propenal |
| CAS | 87424-82-6 |
| Molecular Formula | C10 H10 O3 |
| Quantity | 10 mg |
| Synonym | 2-Propenal','2-methyl-3-(4-methyl-2-oxo-2H-pyran-6-yl)-','(E)- (9CI), (E)-2-Methyl-3-(4-methyl-2-oxo-2H-pyran-6-yl)propenal |
| InChI Formula | InChI=1S/C10H10O3/c1-7-3-9(4-8(2)6-11)13-10(12)5-7/h3-6H,1-2H3/b8-4+ |
| SMILES | C\C(=C/C1=CC(=CC(=O)O1)C)\C=O |
| IUPAC Name | (E)-2-methyl-3-(4-methyl-6-oxopyran-2-yl)prop-2-enal |
| Molecular Weight (g/mol) | 178.18 |
| Formula Weight | 178.063 |