missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ E 2012

Description
E 2012 is a γ-secretase modulator. Lowers Aβ40 and Aβ42 levels and increases Aβ37 levels, in vitro and in vivo. Displays no effect on γ-secretase processing of Notch, EphA4 and EphB2.
Specifications
Specifications
| Chemical Name or Material | (3E)-1-[(1S)-1-(4-Fluorophenyl)ethyl]-3-[[3-methoxy-4-(4-methyl-1H-imidazol-1-yl)phenyl]methylene]-2-piperidone |
| CAS | 870843-42-8 |
| Quantity | 10 mg |
| Molecular Formula | C25H26FN3O2 |
| Formula Weight | 419.49 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?