missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-(Furan-2-yl)acrylaldehyde, TRC

Product Code. 30407864
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407864 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407864 Supplier Toronto Research Chemicals Supplier No. TRCF8402405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39511-08-5
Molecular Formula C7H6O2
Quantity 5 g
Synonym (2E)-3-(2-Furanyl)-2-propenal, (2E)-3-(2-Furyl)acrolein, (2E)-3-(2-Furyl)prop-2-enal, (E)-3-(2-Furanyl)acrolein, (E)-3-(2-Furyl)-2-propenal, (E)-3-(2-Furyl)acrolein, (E)-3-(Furan-2-yl)-2-propenal, (E)-3-(Furan-2-yl)propenal, trans-3-(2-Furanyl)propenal, trans-3-(2-Furyl)-2-propenal, trans-3-(2-Furyl)acrolein, trans-beta-2-Furylacrolein
InChI Formula InChI=1S/C7H6O2/c8-5-1-3-7-4-2-6-9-7/h1-6H/b3-1+
SMILES O=C\C=C\c1occc1
Molecular Weight (g/mol) 122.12
Formula Weight 122.0368
Chemical Name or Material (E)-3-(Furan-2-yl)acrylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.