Learn More
E-4031, MedChemExpress
Product Code. 16472707
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472707 50 mg 50mg
16466618 10 mg 10mg
16429187 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16472707 Supplier Medchem Express Supplier No. HY15551
MedChemExpress E-4031 is a selective hERG potassium channel blocker for use in class III anti-arrhythmic studies.
E-4031 is a selective hERG potassium channel blocker for use in class III anti-arrhythmic studies.
Specifications
Chemical Name or Material E-4031
CAS 113559-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H29Cl2N3O3S
Quantity 50 mg
Solubility Information H2O : ≥ 50 mg/mL (105.39 mM)
SMILES CS(=O)(NC1=CC=C(C(C2CCN(CCC3=NC(C)=CC=C3)CC2)=O)C=C1)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 474.44
Formula Weight 474.44
Percent Purity 96.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only