missing translation for 'onlineSavingsMsg'
Learn More

E-4031, MedChemExpress

Product Code. 16466618
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466618

Brand: Medchem Express HY15551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E-4031 is a selective hERG potassium channel blocker for use in class III anti-arrhythmic studies.

E-4031 is a selective hERG potassium channel blocker for use in class III anti-arrhythmic studies.

Specifications

Chemical Name or Material E-4031
CAS 113559-13-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H29Cl2N3O3S
Quantity 10 mg
Solubility Information H2O : ≥ 50 mg/mL (105.39 mM)
SMILES CS(=O)(NC1=CC=C(C(C2CCN(CCC3=NC(C)=CC=C3)CC2)=O)C=C1)=O.[H]Cl.[H]Cl
Molecular Weight (g/mol) 474.44
Formula Weight 474.44
Percent Purity 96.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.