missing translation for 'onlineSavingsMsg'
Learn More

E-7386, MedChemExpress

Product Code. 16474438
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474438 10 mg 10mg
16420757 50 mg 50mg
16417167 5 mg 5mg
16411159 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474438 Supplier Medchem Express Supplier No. HY111386

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E-7386 is an orally active CBP/beta-catenin modulator.

E-7386 is an orally active CBP/beta-catenin modulator.

Specifications

Chemical Name or Material E-7386
CAS 1799824-08-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C39H48FN9O4
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (172.21 mM; Need ultrasonic)
SMILES O=C(N([C@](CN1CC2=NC(N3CC(N4CCN(CC)CC4)C3)=CC=C2)([H])N5[C@@H](CC6=CC=C(O)C=C6F)C1=O)N(CC=C)CC5=O)NCC7=CC=CC=C7
Molecular Weight (g/mol) 725.85
Formula Weight 725.85
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.